Learn More
Methyl malonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 414510050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl malonyl chloride | |
37517-81-0 | |
1.2730g/mL | |
80°C | |
97% | |
C4H5ClO3 | |
ClCOCH2CO2CH3 | |
5 g | |
1.273 | |
UTBCRHAMJFMIIR-UHFFFAOYSA-N | |
methyl 3-chloro-3-oxopropanoate | |
123460 | |
97% |
97% | |
Yellow | |
57.0°C to 59.0°C (12.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4310 to 1.4350 | |
MFCD00013657 | |
02, IV, 1886 | |
methyl malonyl chloride, methyl 3-chloro-3-oxopropionate, methyl chloroformylacetate, propanoic acid, 3-chloro-3-oxo-, methyl ester, methyl-3-chloro-3-oxopropionate, methyl-3-chlor-3-oxopropanoat, methyl chlorocarbonylacetate, carbomethoxy acetyl chloride, methyl chloroformyl acetate, methyl3-chloro-3-oxopropionate | |
COC(=O)CC(=O)Cl | |
136.53 | |
136.53 | |
Liquid |
Chemical Identifiers
37517-81-0 | |
136.53 | |
UTBCRHAMJFMIIR-UHFFFAOYSA-N | |
methyl 3-chloro-3-oxopropanoate |
C4H5ClO3 | |
MFCD00013657 | |
123460 | |
COC(=O)CC(=O)Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 253-540-6
Recommended Storage : Corrosives
RUO – Research Use Only