missing translation for 'onlineSavingsMsg'
Learn More

Methyl malonyl chloride, 97%

CAS: 37517-81-0 | C4H5ClO3 | 136.53 g/mol

$125.91 - $386.66

Chemical Identifiers

CAS 37517-81-0
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.53
MDL Number MFCD00013657
InChI Key UTBCRHAMJFMIIR-UHFFFAOYSA-N
PubChem CID 123460
IUPAC Name methyl 3-chloro-3-oxopropanoate
SMILES COC(=O)CC(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC414510050
SDS View Documents
Thermo Scientific Chemicals
414510050
5 g Glass bottle
Each for $125.91
Only null left
 
AC414510250
SDS View Documents
Thermo Scientific Chemicals
414510250
25 g Glass bottle
Each for $386.66
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

37517-81-0
136.53
UTBCRHAMJFMIIR-UHFFFAOYSA-N
methyl 3-chloro-3-oxopropanoate
C4H5ClO3
MFCD00013657
123460
COC(=O)CC(=O)Cl

Specifications

37517-81-0
1.2730g/mL
80°C
97%
C4H5ClO3
ClCOCH2CO2CH3
5 g
1.273
UTBCRHAMJFMIIR-UHFFFAOYSA-N
methyl 3-chloro-3-oxopropanoate
123460
97%
Methyl malonyl chloride, 97%
Yellow
57.0°C to 59.0°C (12.0 mmHg)
Authentic
Glass bottle
1.4310 to 1.4350
MFCD00013657
02, IV, 1886
methyl malonyl chloride, methyl 3-chloro-3-oxopropionate, methyl chloroformylacetate, propanoic acid, 3-chloro-3-oxo-, methyl ester, methyl-3-chloro-3-oxopropionate, methyl-3-chlor-3-oxopropanoat, methyl chlorocarbonylacetate, carbomethoxy acetyl chloride, methyl chloroformyl acetate, methyl3-chloro-3-oxopropionate
COC(=O)CC(=O)Cl
136.53
136.53
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 253-540-6

Recommended Storage : Corrosives

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only