missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl octanoate, 99%, analytical standard for GC, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 460350010

 View more versions of this product

Catalog No. AC460350010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl octanoate
0.8780g/mL
73°C
98.5% min. (GC)
C9H18O2
1.4165 to 1.4185
0.878
Solubility in water: insoluble.
CCCCCCCC(=O)OC
158.24
CHEBI:87432
99%
111-11-5
79.0°C (12.0 mmHg)
Authentic
Glass Bottle
CH3(CH2)6COOCH3
1g
methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728
JGHZJRVDZXSNKQ-UHFFFAOYSA-N
methyl octanoate
8091
158.24
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only