missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl octanoate, 99%, analytical standard for GC, Thermo Scientific™
Specifications
CAS | 111-11-5 |
---|---|
Molecular Formula | C9H18O2 |
Molecular Weight (g/mol) | 158.24 |
InChI Key | JGHZJRVDZXSNKQ-UHFFFAOYSA-N |
Synonym | methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC460350010
|
Thermo Scientific Chemicals
460350010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC460350050
|
Thermo Scientific Chemicals
460350050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
111-11-5 | |
158.24 | |
methyl caprylate, caprylic acid methyl ester, methyl n-octanoate, octanoic acid, methyl ester, uniphat a20, octanoic acid methyl ester, methyl octylate, methyl caprylate natural, unii-7mo740x6ql, fema no. 2728 | |
CHEBI:87432 | |
CCCCCCCC(=O)OC |
C9H18O2 | |
JGHZJRVDZXSNKQ-UHFFFAOYSA-N | |
8091 | |
methyl octanoate |
RUO â Research Use Only