Methyl Orange, Reagent, ACS, Spectrum™ Chemical
ME160, 547-58-0, C14H14N3O3SNa
Supplier:  Spectrum Chemical Mfg Cor ME160500GMGL
 View more versions of this product
Catalog No. 18604437
Explore available promotions
Description
Description
Spectrum™ Chemical Methyl Orange, Reagent, ACS is most typically used as an indicator in titrations due to the fact of its color change. The color changes happen in acidic conditions and usually moves from red to orange and eventually to yellow. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications
Specifications
Methyl Orange
100%
UN3143
CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O
304.34
Amber Glass Bottle
500 g
547-58-0
C14H14N3O3S
IETWCRRCPURZOC-UHFFFAOYSA-M
4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
ACS
Orange to Yellow
Chemical Identifiers
547-58-0
304.34
4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
C14H14N3O3S
IETWCRRCPURZOC-UHFFFAOYSA-M
CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O