missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl Orange, Reagent, ACS, Spectrum™ Chemical
ME160, 547-58-0, C14H14N3O3SNa
$238.59 - $8011.26
Chemical Identifiers
CAS | 547-58-0 |
---|---|
Molecular Formula | C14H14N3O3S |
Molecular Weight (g/mol) | 304.34 |
InChI Key | IETWCRRCPURZOC-UHFFFAOYSA-M |
IUPAC Name | 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate |
SMILES | CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18604434
|
Spectrum Chemical Mfg Cor
ME16025GMGL |
25 g | Amber Glass Bottle |
Each for $238.59
|
|
||||
18604435
|
Spectrum Chemical Mfg Cor
ME160125GMGL |
125 g | Amber Glass Bottle |
Each for $518.41
|
|
||||
18604437
|
Spectrum Chemical Mfg Cor
ME160500GMGL |
500 g | Amber Glass Bottle |
Each for $1,429.29
|
|
||||
18604436
|
Spectrum Chemical Mfg Cor
ME160125GMGLCS6 |
6 x 125 g | Amber Glass Bottle |
Case of 6 Each for $2,013.41
|
|
||||
18604438
|
Spectrum Chemical Mfg Cor
ME160500GMGLCS6 |
6 x 500 g | Amber Glass Bottle |
Case of 6 Each for $7,887.38
|
|
||||
18604439
|
Spectrum Chemical Mfg Cor
ME1605KGBL |
5 kg | Poly Pail |
Each for $3,316.53
|
|
||||
18604440
|
Spectrum Chemical Mfg Cor
ME16025KGBL |
25 kg | Fiber Drum |
Each for $8,011.26
|
|
||||
Description
Spectrum™ Chemical Methyl Orange, Reagent, ACS is most typically used as an indicator in titrations due to the fact of its color change. The color changes happen in acidic conditions and usually moves from red to orange and eventually to yellow. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Chemical Identifiers
547-58-0 | |
304.34 | |
4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate |
C14H14N3O3S | |
IETWCRRCPURZOC-UHFFFAOYSA-M | |
CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O |
Specifications
547-58-0 | |
C14H14N3O3S | |
IETWCRRCPURZOC-UHFFFAOYSA-M | |
4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate | |
ACS | |
25 g |
100% | |
UN3143 | |
CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O | |
304.34 | |
Amber Glass Bottle |