missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenyl sulfoxide, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 352120250

 View more versions of this product

Catalog No. AC352120250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl phenyl sulfoxide
1193-82-4
86°C
98% min. (GC)
C7H8OS
C6H5SOCH3
25g
Solubility in water: insoluble (20°C)
CS(=O)C1=CC=CC=C1
140.21
140.21
98%
92°C to 94°C (0.3mmHg)
Authentic
Glass bottle
1.5775
MFCD00002088
methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
JXTGICXCHWMCPM-UHFFFAOYSA-N
methylsulfinylbenzene
14516
98+%

Chemical Identifiers

1193-82-4
140.21
JXTGICXCHWMCPM-UHFFFAOYSA-N
14516
CS(=O)C1=CC=CC=C1
C7H8OS
MFCD00002088
methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
methylsulfinylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

HYGROSCOPIC

EINECSNumber : 214-781-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only