Learn More
Methyl tert-Butyl Ether (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Supplier: Thermo Fisher Scientific FLE1274
Specifications
Methyl tert-Butyl Ether | |
1634-04-4 | |
Colorless | |
54°C | |
−28°C | |
Amber Glass Bottle | |
Mobile phase for HPLC applications | |
10ppm max. | |
C5H12O | |
268mbar at 20°C | |
0.05% max. | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
2-methoxy-2-methylpropane | |
15413 | |
88.14 | |
HPLC |
-110°C | |
0.014 AU max. at 300 nm, 0.10 AU max. at 254 nm, 0.50 AU max. at 225 nm, 0.80 AU max. at 210 nm | |
0.740g/cm³ | |
4 L | |
Pass Test | |
Liquid | |
1.3650 to 1.3670 (at 25°C) | |
99% min by GC | |
MFCD00008812 | |
2398 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
CC(C)(C)OC | |
88.15 | |
CHEBI:27642 | |
≥99% |
Chemical Identifiers
1634-04-4 | |
88.15 | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
15413 | |
2-methoxy-2-methylpropane |
C5H12O | |
MFCD00008812 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
CHEBI:27642 | |
CC(C)(C)OC |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.
NFPA
Health:2
Flammability:3
Instability:0
DOTInformation : Class 3: Flammable Liquid