Learn More
Methyl tert-Butyl Ether (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
$954.00 - $954.00
Chemical Identifiers
CAS | 1634-04-4 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00008812 |
InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
Synonym | tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane |
PubChem CID | 15413 |
ChEBI | CHEBI:27642 |
IUPAC Name | 2-methoxy-2-methylpropane |
SMILES | CC(C)(C)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
E127-4
|
Thermo Fisher Scientific
FLE1274 |
4 L |
Each for $954.00
Case of 4 Each for $3,816.00
|
|
|||||
Chemical Identifiers
1634-04-4 | |
88.15 | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
15413 | |
2-methoxy-2-methylpropane |
C5H12O | |
MFCD00008812 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
CHEBI:27642 | |
CC(C)(C)OC |
Specifications
-110°C | |
0.014 AU max. at 300 nm, 0.10 AU max. at 254 nm, 0.50 AU max. at 225 nm, 0.80 AU max. at 210 nm | |
0.740g/cm³ | |
4 L | |
Pass Test | |
Liquid | |
10ppm max. | |
C5H12O | |
268mbar at 20°C | |
0.05% max. | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
2-methoxy-2-methylpropane | |
15413 | |
88.14 | |
Mobile phase for HPLC applications |
1634-04-4 | |
Colorless | |
54°C | |
−28°C | |
Amber Glass Bottle | |
1.3650 to 1.3670 (at 25°C) | |
99% min by GC | |
MFCD00008812 | |
2398 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
CC(C)(C)OC | |
88.15 | |
CHEBI:27642 | |
≥99% | |
Methyl tert-Butyl Ether |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.
NFPA
Health:2
Flammability:3
Instability:0
DOTInformation : Class 3: Flammable Liquid