Learn More
Methyl thiosalicylate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2840014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Methyl thiosalicylate | |
1.223 | |
>110°C (230°F) | |
1.592 | |
25 g | |
methyl thiosalicylate, methyl 2-mercaptobenzoate, methyl o-mercaptobenzoate, usaf ma-15, benzoic acid, 2-mercapto-, methyl ester, methyl-o-mercaptobenzoate, methyl-2-mercaptobenzoate, benzoic acid, o-mercapto-, methyl ester, 2-methoxycarbonyl thiophenol, 2-mercaptobenzoic acid methyl ester | |
COC(=O)C1=CC=CC=C1S | |
168.21 | |
168.21 |
4892-02-8 | |
98°C to 100°C (2 mmHg) | |
C8H8O2S | |
MFCD00060678 | |
Air Sensitive | |
BAQGCWNPCFABAY-UHFFFAOYSA-N | |
methyl 2-sulfanylbenzoate | |
21009 | |
97% |
Identifiants chimiques
4892-02-8 | |
MFCD00060678 |
Sécurité et manipulation
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only