missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl thiosalicylate, 97%, Thermo Scientific Chemicals

$161.99 - $458.37

Chemical Identifiers

CAS 4892-02-8
MDL Number MFCD00060678
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2840006
View Documents
Thermo Scientific Chemicals
H2840006
5 g
Each for $161.99
 
AAH2840014
View Documents
Thermo Scientific Chemicals
H2840014
25 g
Each for $458.37
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4892-02-8
MFCD00060678

Specifications

4892-02-8
98°C to 100°C (2 mmHg)
C8H7ClO2
MFCD00060678
Air Sensitive
WYVGYYIZXPXHAZ-UHFFFAOYSA-N
3-chloro-4-methoxybenzaldehyde
78619
97%
1.223
>110°C (230°F)
1.592
5 g
3-chloroanisaldehyde, 3-chloro-p-anisaldehyde, benzaldehyde, 3-chloro-4-methoxy, 3-chloro-4-methoxybenzenecarbaldehyde, 3-chloro-4-methoxy-benzaldehyde, pubchem2638, acmc-1ap2w, 3-chloro-4-methoxybenzaldehyde
COC1=C(C=C(C=C1)C=O)Cl
170.59
168.21
Methyl thiosalicylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only