missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl thiosalicylate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2840006

 View more versions of this product

Catalog No. AAH2840006


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl thiosalicylate
1.223
>110°C (230°F)
1.592
5 g
3-chloroanisaldehyde, 3-chloro-p-anisaldehyde, benzaldehyde, 3-chloro-4-methoxy, 3-chloro-4-methoxybenzenecarbaldehyde, 3-chloro-4-methoxy-benzaldehyde, pubchem2638, acmc-1ap2w, 3-chloro-4-methoxybenzaldehyde
COC1=C(C=C(C=C1)C=O)Cl
170.59
168.21
4892-02-8
98°C to 100°C (2 mmHg)
C8H7ClO2
MFCD00060678
Air Sensitive
WYVGYYIZXPXHAZ-UHFFFAOYSA-N
3-chloro-4-methoxybenzaldehyde
78619
97%

Chemical Identifiers

4892-02-8
MFCD00060678
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only