missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle6-morpholinonicotinate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC17522DE

 View more versions of this product

Catalog No. CC17522DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Methyle6-morpholinonicotinate
96.5
97%
MFCD03783347
methyl 6-morpholinonicotinate, methyl 6-morpholin-4-yl pyridine-3-carboxylate, acmc-1by02, methyl 6-morpholin-4-ylnicotinate, methyl6-morpholinonicotinate, methyl 6-morpholin-4-yl-nicotinate, 6-4-morpholinyl-3-pyridinecarboxylic acid methyl ester
COC(=O)C1=CN=C(C=C1)N1CCOCC1
222.24
222.24
132546-81-7
100.0
C11H14N2O3
5g
FFBPRHFKXVNLDV-UHFFFAOYSA-N
methyl 6-morpholin-4-ylpyridine-3-carboxylate
2776466
97%

Chemical Identifiers

132546-81-7
222.24
FFBPRHFKXVNLDV-UHFFFAOYSA-N
2776466
COC(=O)C1=CN=C(C=C1)N1CCOCC1
C11H14N2O3
MFCD03783347
methyl 6-morpholinonicotinate, methyl 6-morpholin-4-yl pyridine-3-carboxylate, acmc-1by02, methyl 6-morpholin-4-ylnicotinate, methyl6-morpholinonicotinate, methyl 6-morpholin-4-yl-nicotinate, 6-4-morpholinyl-3-pyridinecarboxylic acid methyl ester
methyl 6-morpholin-4-ylpyridine-3-carboxylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions