missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyle6-morpholinonicotinate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 132546-81-7
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.24
MDL Number MFCD03783347
InChI Key FFBPRHFKXVNLDV-UHFFFAOYSA-N
Synonym methyl 6-morpholinonicotinate, methyl 6-morpholin-4-yl pyridine-3-carboxylate, acmc-1by02, methyl 6-morpholin-4-ylnicotinate, methyl6-morpholinonicotinate, methyl 6-morpholin-4-yl-nicotinate, 6-4-morpholinyl-3-pyridinecarboxylic acid methyl ester
PubChem CID 2776466
IUPAC Name methyl 6-morpholin-4-ylpyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)N1CCOCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC17522DA
SDS View Documents
Thermo Scientific Chemicals
CC17522DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC17522DE
SDS View Documents
Thermo Scientific Chemicals
CC17522DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

132546-81-7
222.24
FFBPRHFKXVNLDV-UHFFFAOYSA-N
2776466
COC(=O)C1=CN=C(C=C1)N1CCOCC1
C11H14N2O3
MFCD03783347
methyl 6-morpholinonicotinate, methyl 6-morpholin-4-yl pyridine-3-carboxylate, acmc-1by02, methyl 6-morpholin-4-ylnicotinate, methyl6-morpholinonicotinate, methyl 6-morpholin-4-yl-nicotinate, 6-4-morpholinyl-3-pyridinecarboxylic acid methyl ester
methyl 6-morpholin-4-ylpyridine-3-carboxylate

Specifications

132546-81-7
100.0
Amber glass bottle
MFCD03783347
methyl 6-morpholinonicotinate, methyl 6-morpholin-4-yl pyridine-3-carboxylate, acmc-1by02, methyl 6-morpholin-4-ylnicotinate, methyl6-morpholinonicotinate, methyl 6-morpholin-4-yl-nicotinate, 6-4-morpholinyl-3-pyridinecarboxylic acid methyl ester
COC(=O)C1=CN=C(C=C1)N1CCOCC1
222.24
222.24
Methyle6-morpholinonicotinate
96.5
97%
C11H14N2O3
1g
FFBPRHFKXVNLDV-UHFFFAOYSA-N
methyl 6-morpholin-4-ylpyridine-3-carboxylate
2776466
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'