missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylene di-p-phenyl diisocyanate, 98%, flakes, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 414285000

 View more versions of this product

Catalog No. AC414285000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methylene di-p-phenyl diisocyanate
101-68-8,5873-54-1
White
392.0°C
Authentic
Glass bottle
OCNC6H4CH2C6H4NCO
500 g
4,4'-diphenylmethane diisocyanate, diphenylmethane diisocyanate, isonate, 4,4'-diisocyanatodiphenylmethane, p,p'-diphenylmethane diisocyanate, bis 4-isocyanatophenyl methane, 4,4'-methylenebis phenyl isocyanate, methylbisphenyl isocyanate, 1,1'-methylenebis 4-isocyanatobenzene, methylene diphenyl diisocyanate
UPMLOUAZCHDJJD-UHFFFAOYSA-N
1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene
7570
CHEBI:53218
98%
99.5%
40.0°C
1.2200g/mL
202°C
2.5% max. (2,4′-MDI isomer) (GC)
C15H10N2O2
MFCD00036131
1.22
Solubility in water: decomposes. Other solubilities: soluble in acetone,benzene,nitrobenzene
C1=CC(=CC=C1CC2=CC=C(C=C2)N=C=O)N=C=O
250.26
3-6 mPa.s (42°C)
250.26
Flakes

Chemical Identifiers

101-68-8
250.26
UPMLOUAZCHDJJD-UHFFFAOYSA-N
7570
1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene
C15H10N2O2
MFCD00036131
4,4'-diphenylmethane diisocyanate, diphenylmethane diisocyanate, isonate, 4,4'-diisocyanatodiphenylmethane, p,p'-diphenylmethane diisocyanate, bis 4-isocyanatophenyl methane, 4,4'-methylenebis phenyl isocyanate, methylbisphenyl isocyanate, 1,1'-methylenebis 4-isocyanatobenzene, methylene diphenyl diisocyanate
CHEBI:53218
C1=CC(=CC=C1CC2=CC=C(C=C2)N=C=O)N=C=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Harmf

GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g

GHS Signal Word: Danger

EINECSNumber : 202-966-0

RTECSNumber : NQ9350000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only