missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylene di-p-phenyl diisocyanate, 98%, flakes, Thermo Scientific Chemicals

$41.06 - $138.14

Chemical Identifiers

CAS 101-68-8
Molecular Formula C15H10N2O2
Molecular Weight (g/mol) 250.26
MDL Number MFCD00036131
InChI Key UPMLOUAZCHDJJD-UHFFFAOYSA-N
Synonym 4,4'-diphenylmethane diisocyanate, diphenylmethane diisocyanate, isonate, 4,4'-diisocyanatodiphenylmethane, p,p'-diphenylmethane diisocyanate, bis 4-isocyanatophenyl methane, 4,4'-methylenebis phenyl isocyanate, methylbisphenyl isocyanate, 1,1'-methylenebis 4-isocyanatobenzene, methylene diphenyl diisocyanate
PubChem CID 7570
ChEBI CHEBI:53218
IUPAC Name 1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene
SMILES C1=CC(=CC=C1CC2=CC=C(C=C2)N=C=O)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC414281000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
414281000
100 g Glass bottle
Each for $41.06
 
AC414285000
SDS View Documents Promotion Details
Thermo Scientific Chemicals
414285000
500 g Glass bottle
Each for $138.14
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

101-68-8
250.26
UPMLOUAZCHDJJD-UHFFFAOYSA-N
7570
1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene
C15H10N2O2
MFCD00036131
4,4'-diphenylmethane diisocyanate, diphenylmethane diisocyanate, isonate, 4,4'-diisocyanatodiphenylmethane, p,p'-diphenylmethane diisocyanate, bis 4-isocyanatophenyl methane, 4,4'-methylenebis phenyl isocyanate, methylbisphenyl isocyanate, 1,1'-methylenebis 4-isocyanatobenzene, methylene diphenyl diisocyanate
CHEBI:53218
C1=CC(=CC=C1CC2=CC=C(C=C2)N=C=O)N=C=O

Specifications

101-68-8,5873-54-1
White
392.0°C
Authentic
Glass bottle
OCNC6H4CH2C6H4NCO
100 g
4,4'-diphenylmethane diisocyanate, diphenylmethane diisocyanate, isonate, 4,4'-diisocyanatodiphenylmethane, p,p'-diphenylmethane diisocyanate, bis 4-isocyanatophenyl methane, 4,4'-methylenebis phenyl isocyanate, methylbisphenyl isocyanate, 1,1'-methylenebis 4-isocyanatobenzene, methylene diphenyl diisocyanate
UPMLOUAZCHDJJD-UHFFFAOYSA-N
1-isocyanato-4-[(4-isocyanatophenyl)methyl]benzene
7570
CHEBI:53218
98%
Methylene di-p-phenyl diisocyanate, 99.5%
40.0°C
1.2200g/mL
202°C
2.5% max. (2,4′-MDI isomer) (GC)
C15H10N2O2
MFCD00036131
1.22
Solubility in water: decomposes. Other solubilities: soluble in acetone,benzene,nitrobenzene
C1=CC(=CC=C1CC2=CC=C(C=C2)N=C=O)N=C=O
250.26
3-6 mPa.s (42°C)
250.26
Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Harmf

GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective g

GHS Signal Word: Danger

EINECSNumber : 202-966-0

RTECSNumber : NQ9350000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only