missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methylhydrazine Sulfate 98.0+%, TCI America™
Supplier: TCI America M03415G
Specifications
Methylhydrazine Sulfate | |
143°C | |
CH8N2O4S | |
5 g | |
methylhydrazine sulfate, methylhydrazine; sulfuric acid, methyl hydrazine sulfate, methyl hydrazine sulphate, hydrazine, methyl-, sulfate, methyl hydrazine; sulfuric acid, ch6n2.h2o4s, methylhydrazine sulphate, dsstox_cid_875, n-methyl hydrazine sulfate | |
CN[NH3+].OS([O-])(=O)=O | |
144.15 | |
144.15 | |
Crystalline Powder |
302-15-8 | |
White-Yellow | |
MFCD00035423 | |
2811 | |
KJDJPXUIZYHXEZ-UHFFFAOYSA-N | |
2-methylhydrazinium hydrogen sulfate | |
165286 | |
≥98.0% (T) |
Chemical Identifiers
302-15-8 | |
144.15 | |
KJDJPXUIZYHXEZ-UHFFFAOYSA-N | |
165286 | |
CN[NH3+].OS([O-])(=O)=O |
CH8N2O4S | |
MFCD00035423 | |
methylhydrazine sulfate, methylhydrazine; sulfuric acid, methyl hydrazine sulfate, methyl hydrazine sulphate, hydrazine, methyl-, sulfate, methyl hydrazine; sulfuric acid, ch6n2.h2o4s, methylhydrazine sulphate, dsstox_cid_875, n-methyl hydrazine sulfate | |
2-methylhydrazinium hydrogen sulfate |
Safety and Handling
RTECSNumber : MV7750000
TSCA : Yes