missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methylhydrazine Sulfate 98.0+%, TCI America™
$108.62 - $353.52
Chemical Identifiers
CAS | 302-15-8 |
---|---|
Molecular Formula | CH8N2O4S |
Molecular Weight (g/mol) | 144.15 |
MDL Number | MFCD00035423 |
InChI Key | KJDJPXUIZYHXEZ-UHFFFAOYSA-N |
Synonym | methylhydrazine sulfate, methylhydrazine; sulfuric acid, methyl hydrazine sulfate, methyl hydrazine sulphate, hydrazine, methyl-, sulfate, methyl hydrazine; sulfuric acid, ch6n2.h2o4s, methylhydrazine sulphate, dsstox_cid_875, n-methyl hydrazine sulfate |
PubChem CID | 165286 |
IUPAC Name | 2-methylhydrazinium hydrogen sulfate |
SMILES | CN[NH3+].OS([O-])(=O)=O |
Chemical Identifiers
302-15-8 | |
144.15 | |
KJDJPXUIZYHXEZ-UHFFFAOYSA-N | |
165286 | |
CN[NH3+].OS([O-])(=O)=O |
CH8N2O4S | |
MFCD00035423 | |
methylhydrazine sulfate, methylhydrazine; sulfuric acid, methyl hydrazine sulfate, methyl hydrazine sulphate, hydrazine, methyl-, sulfate, methyl hydrazine; sulfuric acid, ch6n2.h2o4s, methylhydrazine sulphate, dsstox_cid_875, n-methyl hydrazine sulfate | |
2-methylhydrazinium hydrogen sulfate |
Specifications
302-15-8 | |
White-Yellow | |
MFCD00035423 | |
2811 | |
KJDJPXUIZYHXEZ-UHFFFAOYSA-N | |
2-methylhydrazinium hydrogen sulfate | |
165286 | |
≥98.0% (T) | |
Methylhydrazine Sulfate |
143°C | |
CH8N2O4S | |
5 g | |
methylhydrazine sulfate, methylhydrazine; sulfuric acid, methyl hydrazine sulfate, methyl hydrazine sulphate, hydrazine, methyl-, sulfate, methyl hydrazine; sulfuric acid, ch6n2.h2o4s, methylhydrazine sulphate, dsstox_cid_875, n-methyl hydrazine sulfate | |
CN[NH3+].OS([O-])(=O)=O | |
144.15 | |
144.15 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : MV7750000
TSCA : Yes