missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylmalonic acid, 96%
CAS: 516-05-2 | C4H6O4 | 118.09 g/mol
Supplier: Thermo Scientific Chemicals 127091000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methylmalonic acid | |
| 95.0 | |
| 129.0°C to 136.0°C | |
| 1.4500g/mL | |
| 96% | |
| C4H6O4 | |
| MFCD00002656 | |
| 02, IV, 1932 | |
| methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7 | |
| ZIYVHBGGAOATLY-UHFFFAOYSA-N | |
| 2-methylpropanedioic acid | |
| 487 | |
| 118.09 | |
| Crystalline Powder |
| 516-05-2 | |
| 100.0 | |
| White | |
| Authentic | |
| Glass bottle | |
| HO2CCH(CH3)CO2H | |
| 100 g | |
| 1.45 | |
| Solubility in water: very soluble. Other solubilities: very soluble in ethanol,soluble in ether and ethyl acetate,slightly soluble in benzene and trifluoroacetic,acid | |
| CC(C(O)=O)C(O)=O | |
| 118.09 | |
| CHEBI:30860 | |
| 96% |
Chemical Identifiers
| 516-05-2 | |
| 118.09 | |
| ZIYVHBGGAOATLY-UHFFFAOYSA-N | |
| 487 | |
| 2-methylpropanedioic acid |
| C4H6O4 | |
| MFCD00002656 | |
| methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7 | |
| CHEBI:30860 | |
| CC(C(O)=O)C(O)=O |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 208-219-5
RUO – Research Use Only