missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylmalonic acid, 96%, Thermo Scientific Chemicals

$1226.57 - $1226.57

Chemical Identifiers

CAS 516-05-2
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002656
InChI Key ZIYVHBGGAOATLY-UHFFFAOYSA-N
Synonym methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7
PubChem CID 487
ChEBI CHEBI:30860
IUPAC Name 2-methylpropanedioic acid
SMILES CC(C(O)=O)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC127091000
SDS View Documents
Thermo Scientific Chemicals
127091000
100 g Glass bottle
Each for $1,226.57
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

516-05-2
118.09
ZIYVHBGGAOATLY-UHFFFAOYSA-N
487
2-methylpropanedioic acid
C4H6O4
MFCD00002656
methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7
CHEBI:30860
CC(C(O)=O)C(O)=O

Specifications

516-05-2
100.0
White
Authentic
Glass bottle
HO2CCH(CH3)CO2H
100 g
1.45
Solubility in water: very soluble. Other solubilities: very soluble in ethanol,soluble in ether and ethyl acetate,slightly soluble in benzene and trifluoroacetic,acid
CC(C(O)=O)C(O)=O
118.09
CHEBI:30860
96%
Methylmalonic acid
95.0
129.0°C to 136.0°C
1.4500g/mL
96%
C4H6O4
MFCD00002656
02, IV, 1932
methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7
ZIYVHBGGAOATLY-UHFFFAOYSA-N
2-methylpropanedioic acid
487
118.09
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 208-219-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only