missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylparaben, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor H22655KG

 View more versions of this product

Catalog No. 18611806

Explore available promotions

Description

Description

Specifications

Specifications

99-76-3
270 to 280°C
5 kg
Crystalline Powder or Lumps
1
Poly Bottle
Reagent

Chemical Identifiers

99-76-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.