missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methylparaben, Spectrum™ Chemical
$188.24 - $9250.38
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18611805
|
Spectrum Chemical Mfg Cor
H2265100GM |
100 g | Poly Bottle |
Each for $188.24
|
|
|||||
18611808
|
Spectrum Chemical Mfg Cor
H2265500GM |
500 g | Poly Bottle |
Each for $372.18
|
|
|||||
18611806
|
Spectrum Chemical Mfg Cor
H22655KG |
5 kg | Poly Bottle |
Each for $2,694.15
|
|
|||||
18611807
|
Spectrum Chemical Mfg Cor
H226550KGBL |
50 kg | Poly Bottle |
Each for $9,250.38
|
|
|||||
Chemical Identifiers
99-76-3 |
Specifications
99-76-3 | |
270 to 280°C | |
100 g | |
Crystalline Powder or Lumps |
1 | |
Poly Bottle | |
Reagent |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.