missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylparaben, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor H226550KGBL

 View more versions of this product

Catalog No. 18611807


Explore available promotions

Description

Description

Specifications

Specifications

99-76-3
270 to 280°C
50 kg
Crystalline Powder or Lumps
1
Poly Bottle
Reagent

Chemical Identifiers

99-76-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.