missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

Troemner™ Metric Economical Individual Calibration Weights, Class 7

Individual weights are available from 2kg to 1mg

 Afficher plus de versions de ce produit

Numéro de catalogue. 02-306Q


Découvrez plus d'offres spéciales

Description

Description

Weights are adjusted to tolerances specified by ANSI/ASTM E617 Class 7

  • Weights 1g and above are carefully manufactured of stainless steel and are cylindrical in shape for easy gripping
  • Weights 1g to 100g are made from one-piece construction and are adjusted during manufacture by removing material from the bottom of the weight
  • Weights 200g through 2kg have an adjusting cavity on the top that is sealed with a plug
  • Economical stainless steel weights 1g and above are perfectly flat on top to stack easily on center
  • Weights 500mg and lower are made of sheet metal and feature one-piece construction with one side turned up to make them easy to handle with forceps
  • he 500mg and 200mg weights are made of stainless steel while weights 100mg and below are made of aluminum
Spécifications

Spécifications

Individual Metric Economical Weight
Aluminum
2.7g/cm3 at 20°C
Flat with one side turned up
1
ASTM Class 7
100 mg
100 mg
Each
Videos
FDS
Documents

Documents

Offres spéciales

Offres spéciales