Troemner™ Metric Economical Individual Calibration Weights, Class 7
Individual weights are available from 2kg to 1mg
$49.55 - $421.03
Specifications
Product Type Individual Metric Economical Weight
Classification ASTM Class 7
Quantity Each
View More Specs
Products 9
Catalog Number Mfr. No. Mass Density Material Shape Price Quantity  
Catalog Number Mfr. No. Mass Density Material Shape Price Quantity  
02-301-127
View Documents
N/A 500 g 7.85g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
Each for $125.03
02301114
View Documents
Troemner™
30391000
1000 g 7.85g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
Each for $182.32
02-301-111
View Documents
N/A 1 g 7.85g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
Each for $56.66
02-301-120
View Documents
N/A 200 g 7.85g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
Each for $82.28
02-301-113
View Documents
N/A 100 g 7.85g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
Each for $55.76
02-301-118
View Documents
N/A 2 g 7.85g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
Each for $53.38
02-301-121
View Documents
N/A 2000 g 7.85g/cm3 at 20°C Stainless Steel Cylindrical with Handling Knob
Each for $421.03
02-301-122
missing translation for 'viewDocuments'
N/A 5 mg 2.7g/cm3 at 20°C Aluminum Flat with one side turned up
Each for $49.55
02-306Q
missing translation for 'viewDocuments'
N/A 100 mg 2.7g/cm3 at 20°C Aluminum Flat with one side turned up
Each for $51.21
Description
Description
Weights are adjusted to tolerances specified by ANSI/ASTM E617 Class 7
  • Weights 1g and above are carefully manufactured of stainless steel and are cylindrical in shape for easy gripping
  • Weights 1g to 100g are made from one-piece construction and are adjusted during manufacture by removing material from the bottom of the weight
  • Weights 200g through 2kg have an adjusting cavity on the top that is sealed with a plug
  • Economical stainless steel weights 1g and above are perfectly flat on top to stack easily on center
  • Weights 500mg and lower are made of sheet metal and feature one-piece construction with one side turned up to make them easy to handle with forceps
  • he 500mg and 200mg weights are made of stainless steel while weights 100mg and below are made of aluminum
Specifications
Specifications
Individual Metric Economical Weight
ASTM Class 7
Each