Mettler Toledo™ Aluminum Weigh Boats for Micro and Ultramicro Series
For discontinued Micro (MT5) and Ultramicro (UMT2) Series balances
Supplier:  Mettler Toledo™ 110501
 View more versions of this product
Catalog No. 01-910-71
Explore available promotions
Description
Description
For discontinued Micro (MT5) and Ultramicro (UMT2) Series balances
Sturdy yet lightweight boats for sample handling.
Specifications
Specifications
Flat
Rectangular
Discontinued Micro (MT5) and Ultramicro (UMT2) Series Balances
0.12 in.
0.5 in.
4 mm
6 mg
Aluminum
Weighing Boat
0.12 x 0.5 x 0.18 in. (3 x 12.7 x 4.5 mm)
3 mm
12.7 mm
0.18 in.
Product Suggestions
Product Suggestions