missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ Analytical Individual Weights: ASTM™ Class 2
Low magnetic permeability and high corrosion resistance for reliable stable values
$101.86 - $1345.18
Specifications
Classification | ASTM Class 2 |
---|---|
Material | Stainless Steel |
Density | 7.9kg/dm3 at 20°C |
Includes | Plastic box; Without certificate |
Quantity | Each |
Catalog Number | Mfr. No. | Mass | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Mass | Price | Quantity | |||||
01-915-725
|
Mettler Toledo™
11123492 |
1 mg |
Each for $105.28
|
|
|||||
01-915-726
|
Mettler Toledo™
11123493 |
2 mg |
Each for $101.86
|
|
|||||
01-915-727
|
Mettler Toledo™
11123494 |
5 mg |
Each for $101.86
|
|
|||||
01-915-728
|
Mettler Toledo™
11123495 |
10 mg |
Each for $101.86
|
|
|||||
01-915-729
|
Mettler Toledo™
11123496 |
20 mg |
Each for $101.86
|
|
|||||
01-915-731
|
Mettler Toledo™
11123498 |
100 mg |
Each for $107.23
|
|
|||||
01-915-732
|
Mettler Toledo™
11123500 |
500 mg |
Each for $107.23
|
|
|||||
01-915-730
|
Mettler Toledo™
11123497 |
50 mg |
Each for $107.23
|
|
|||||
01-915-734
|
Mettler Toledo™
11123502 |
2 g |
Each for $154.02
|
|
|||||
01-915-733
|
Mettler Toledo™
11123501 |
1 g |
Each for $145.72
|
|
|||||
01-915-735
|
Mettler Toledo™
11123503 |
5 g |
Each for $154.02
|
|
|||||
01-915-736
|
Mettler Toledo™
11123504 |
10 g |
Each for $170.09
|
|
|||||
01-915-737
|
Mettler Toledo™
11123505 |
20 g |
Each for $170.09
|
|
|||||
01-915-738
|
Mettler Toledo™
11123506 |
50 g |
Each for $186.18
|
|
|||||
01-915-743
|
Mettler Toledo™
11123509 |
500 g |
Each for $424.03
|
|
|||||
01-915-740
|
Mettler Toledo™
11123507 |
100 g |
Each for $211.52
|
|
|||||
01-915-742
|
Mettler Toledo™
11123508 |
200 g |
Each for $298.28
|
|
|||||
01-915-745
|
Mettler Toledo™
11123511 |
2 kg |
Each for $1,345.18
|
|
|||||
01-915-744
|
Mettler Toledo™
11123510 |
1 kg |
Each for $901.66
|
|
|||||
Description
Features
- Premium stainless steel
- State-of-the-art turning procedures, improved coatings, stronger substrates
- Electrolytic polishing ensures smooth, glossy surface
- Magnetization and susceptibility of all weights are strictly controlled to ensure compliance with standards
- Two-piece screw-knob construction
- 10mg to 20mg weights are sheet weights made of stainless steel
- 200 to 500mg weights are sheet weights made of steel
- 1g to 1kg weights are cylindrical knob weights made of steel
- Density: 7.9kg/dm3
Specifications
ASTM Class 2 | |
7.9kg/dm3 at 20°C | |
Each |
Stainless Steel | |
Plastic box; Without certificate | |
1 |