Mettler Toledo™ Antistatic Systems
Antistatic systems for discharging electrostatically charged samples prior to weighing. Mettler Toledo Antistatic Systems use a high voltage to generate positively and negatively charged ions which are attracted by the electrostatically charged object and neutralize the disruptive charge on the surface of the weighing sample. As a result, the forces that distort the weighing result also disappear.
Supplier: Mettler Toledo™ 63052302
Description
U-shaped Ionizer
- Can be used with nearly all balances and weighing substances
- Weighing sample or container is discharged by being passed through the U-shaped ionizer when the balance is loaded; sample is neutralized in seconds
- Weighing pan remains accessible
- No drafts in weighing chamber
- Some models include safety warning light that will alert you if the power supply is shorted or faulty in some way
Specifications
Mettler Toledo Balance | |
18 cm | |
27 cm | |
U-shaped Ionizer |
7.6 in. | |
10.62 in. | |
High voltage power pack, Warning light |