Learn More
Mettler Toledo™ Antistatic Systems
Antistatic systems for discharging electrostatically charged samples prior to weighing. Mettler Toledo Antistatic Systems use a high voltage to generate positively and negatively charged ions which are attracted by the electrostatically charged object and neutralize the disruptive charge on the surface of the weighing sample. As a result, the forces that distort the weighing result also disappear.
$3375.14 - $5623.20
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Includes | Price | Quantity | ||||
01-910-021
|
Mettler Toledo™
63052302 |
U-shaped Ionizer | High voltage power pack, Warning light |
Each for $5,623.20
|
|
||||
01-910-022
|
Mettler Toledo™
63052303 |
U-shaped Ionizer | High voltage power pack |
Each for $4,368.18
|
|
||||
09-320-40
|
Mettler Toledo™
63052304 |
One Point Ionizer | High voltage power pack, Stand, Warning light |
Each for $4,916.49
|
|
||||
09-320-41
|
Mettler Toledo™
63052305 |
One Point Ionizer | High voltage power pack |
Each for $3,375.14
|
|
||||
Description
Use a high voltage to generate positively and negatively charged ions. Ions are attracted by the electrostatically charged object and neutralize the disruptive charge on the surface of the weighing sample. As a result, the forces that distort the weighing result also disappear.
U-shaped Ionizer
- Can be used with nearly all balances and weighing substances
- Weighing sample or container is discharged by being passed through the U-shaped ionizer when the balance is loaded; sample is neutralized in seconds
- Weighing pan remains accessible
- No drafts in weighing chamber
- Ideal for use with powdery, toxic samples
- Discharges even the smallest objects
- Probe easily inserted into containers—flexible cable enables fastening in any position simply and easily
Specifications
Mettler Toledo Balance |