Mettler Toledo™ Antistatic Systems
Antistatic systems for discharging electrostatically charged samples prior to weighing. Mettler Toledo Antistatic Systems use a high voltage to generate positively and negatively charged ions which are attracted by the electrostatically charged object and neutralize the disruptive charge on the surface of the weighing sample. As a result, the forces that distort the weighing result also disappear.
Supplier: Mettler Toledo™ 63052304
Description
Point Ionizer
- Ideal for use with powdery, toxic samples
- Discharges even the smallest objects
- Probe easily inserted into containers—flexible cable enables fastening in any position simply and easily
- Some models include safety warning light that will alert you if the power supply is shorted or faulty in some way
Specifications
Mettler Toledo Balance | |
High voltage power pack, Stand, Warning light | |
One Point Ionizer |