missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mettler Toledo™ Redox Buffer Solutions
Storage and refill solutions for redox electrodes.
Supplier: Mettler Toledo™ 51350060
Description
Includes a quality inspection certificate which guarantees the stated values and traceability.Certifications
NIST traceable
Specifications
∼100°C | |
Buffer Solution | |
Quality inspection certificate | |
250 mL Bottle | |
Completely Miscible |
Clear, Yellow | |
Redox Buffer; UH = 427mV | |
220 mV | |
7 |