Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Mettler Toledo™ CarePacs™
For routine testing of balances
$688.54 - $2553.34
Spécifications
Certifications/Compliance ISO9001, ISO14001 environmental standard
Material Stainless Steel
Quantity 2/Set
voir les spécifications
Avertissement:
Requirements for routine testing depend on many factors with process tolerances, associated risks and laboratory practices being the most important ones. Testing should always be performed at (or close to) maximum load and at approximately 5% of the weighing range, or according to regulatory requirements.
Produits 12
Numéro de catalogue Numéro du manufacturier. Mass Classification Product Type Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Mass Classification Product Type Prix Quantité  
01-912-100
Afficher les documents
Mettler Toledo™
11123100
200 g, 20 g ASTM Class 1 Small Weight Set
chaque for $813.94
01-912-102
Afficher les documents
Mettler Toledo™
11123101
10 g & 200 g ASTM Class 1 Small Weight Set
chaque for $813.94
01-912-103
Afficher les documents
Mettler Toledo™
30550614
10 g & 200 g OIML Class F1/F2 Small Weight Set
chaque for $894.15
01-912-104
Afficher les documents
Mettler Toledo™
11123102
5 g & 100 g ASTM Class 1 Small Weight Set
chaque for $701.83
01-912-105
Afficher les documents
Mettler Toledo™
30550615
5 g & 100 g OIML Class E2/F2 Small Weight Set
chaque for $784.69
01-912-106
Afficher les documents
Mettler Toledo™
11123103
2 g & 50 g ASTM Class 1 Small Weight Set
chaque for $688.54
01-912-108
Afficher les documents
Mettler Toledo™
11123107
20 g & 500 g ASTM Class 1 Medium Weight Set
chaque for $989.78
01-912-109
Afficher les documents
Mettler Toledo™
30550649
20 g & 500 g OIML Class F1/F2 Medium Weight Set
chaque for $1,179.48
01-912-110
Afficher les documents
Mettler Toledo™
11123108
50 g & 1 kg ASTM Class 1 Medium Weight Set
chaque for $1,476.77
01-912-112
Afficher les documents
Mettler Toledo™
11123109
100 g & 2000 g ASTM Class 1 Medium Weight Set
chaque for $1,992.00
01-912-114
Afficher les documents
Mettler Toledo™
11123110
200 g & 2 kg ASTM Class 4 Medium Weight Set
chaque for $2,218.63
09-320-120
Afficher les documents
Mettler Toledo™
11123111
200 g & 5 kg ASTM Class 4 Large Weight Set
chaque for $2,553.34
Description
Description
Perform testing by using only two test weights (included). Save time and money, and rely on accurate measuring results; a unique approach to early detection of out-of-process tolerances.
  • Convenient, cost-effective way to limit the risk of working outside of specified process tolerances
  • Tests are performed in the upper and lower weighing range, typically using just two weights SOP's from Mettler Toledo give clear guidance as to how to perform testing reliably
  • Weight class and nominal value of weights are defined according to characteristics of each balance model
  • Comes with ergonomic tweezers, weight forks, cleanroom-approved gloves and cleaning cloths
  • Includes two Mettler Toledo-recommended, balance model-specific test weights
Benefits of CarePacs
  • Ensures accurate and safe operation of balances
  • Lower cost for initial calibration and recalibration compared with traditional weight sets
  • Validation of process tolerances as low as 0.03%
  • Gap between calibrations performed by a qualified service technician is monitored with the correct weights
  • Option to add third weight between 1mg and 100g for customer-specific testing
Spécifications
Spécifications
ISO9001, ISO14001 environmental standard
Stainless Steel
2/Set
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'