METTLER TOLEDO™ InLab™ Conductivity Benchtop Probes
A selection of conductivity probes suitable for all applications.
Supplier:  METTLER TOLEDO™ 30014094
 View more versions of this product
Catalog No. 01-915-112
Explore available promotions
Description
Description
  • Required cables included for METTLER TOLEDO SevenExcellence or SevenCompact
  • Mini-DIN connects to benchtop meter; LTW connects to portable meter
  • Integrated temperature probes
  • Probes with nominal cell constants require calibration before use
  • Probes with certified cell constants are ready to use after verification step - do not calibrate
Accessories & Care
  • Store dry
  • Handle with care to avoid changing the cell constant (cell constant is a physical value)
  • Nominal cell constants require calibration with one standard (see list of standards below)
  • Certified cell constants require verification check with one standard (see list of standards below)
Standards
  • 10 μS/cm, one-time use sachets (30111141)
  • 84 μS/cm, one-time use sachets (30111140)
  • 1413 μS/cm, one-time use sachets (51302049)
  • 12.88 mS/cm, one-time use sachets (51302050)
Specifications
Specifications
0°C to 100°C
General conductivity measurements
Integrated Temperature Sensor
4.72 in.
120 mm
Stainless Steel
Conductivity
InLab™ 741-ISM
0.105 cm-1 certified cell constant
S230 SevenCompact conductivity meter
Mini-DIN
47.22 in.
1.2 m
0.001 to 500 μS/cm
NTC 30kilohm