missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLER TOLEDO™ InLab™ Viscous pH Electrodes
A glass pH electrode with an integrated temperature sensor featuring the SteadyForce™ reference system, which is pressurized to prevent the junction from clogging.
Supplier: METTLER TOLEDO™ 51343150
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Cosmetics
- Gels, soaps, shampoos
- Resins
- Paints
- Varnishes and glues
- Colorants and dyes
- Oily samples
- TRIS compatible
Accessories & Care
- Ceramic Junction and non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
pH Electrode InLab Viscous | |
Ceramic | |
0 to 14 | |
Combination Electrode | |
Highly viscous, highly glutinous samples | |
SteadyForce | |
InLab™ Viscous | |
32°F to 266°F | |
1.57 in. | |
0.23 in. |
S7 | |
Glass | |
0°C to 130°C | |
FRISCOLYT-B | |
S7 to BNC (30281915) | |
HA Glass | |
3 mol/L KCl | |
0°C to 100°C | |
40 mm | |
6 mm |