Mettler Toledo™ Ion-Selective Combination Electrodes: Filling Solutions
For use with ISE combination electrodes.
Supplier:  Mettler Toledo™ 51340030
 View more versions of this product
Catalog No. 01-913-231
Explore available promotions
Description
Description
For use with ISE combination electrodes.
Specifications
Specifications
Electrolyte for Chloride ISE, 20mL
Ion-Selective Combination Electrodes Filling Solutions
Determine the Chloride concentration in samples
32°F to 176°F
20 mL
ISE Combination Electrode
1 mol/L KNO3
20 mL
0°C to 80°C