Mettler Toledo™ Ion-Selective Combination Electrodes: Filling Solutions
For use with ISE combination electrodes.
Supplier: Mettler Toledo™ 51340034
Description
For use with ISE combination electrodes.Specifications
Electrolyte for Nitrate ISE, 20mL | |
Ion-Selective Combination Electrodes Filling Solutions | |
Determine the Nitrate concentration in samples | |
32°F to 122°F | |
20 mL |
ISE Combination Electrode | |
0.5 mol/L MgSO4 | |
20 mL | |
0°C to 50°C |