Mettler Toledo™ Ion-Selective Combination Electrodes: Filling Solutions
For use with ISE combination electrodes.
Supplier:  Mettler Toledo™ 51340033
 View more versions of this product
Catalog No. 01-913-236
Explore available promotions
Description
Description
For use with ISE combination electrodes.
Specifications
Specifications
Electrolyte for Potassium ISE, 20mL
Ion-Selective Combination Electrodes Filling Solutions
Determine the Potassium concentration in samples
32°F to 122°F
20 mL
ISE Combination Electrode
0.5 mol/L MgSO4
20 mL
0°C to 50°C