Mettler Toledo™ Moisture Analyzer Sample Pans
Mettler Toledo sample pans are the only sample pans that are officially supported by Mettler Toledo Moisture Analyzers for optimal use and guaranteed performance.
Supplier:  Mettler Toledo™ 00214462
 View more versions of this product
Catalog No. 01-909-427
Explore available promotions
Description
Description
Choice of washable, reusable stainless chrome-nickel steel pans for halogen moisture analyzers, or aluminum or stainless-steel pans for all Mettler Toledo moisture analyzers.
  • 3.75 in. (96mm) in diameter
Specifications
Specifications
Reusable Pans - Pk of 3
Round
All Mettler Toledo Moisture Analyzers
88.9 mm
Stainless Steel
Moisture Pan
3 Pack