missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLER TOLEDO™ InLab™ Redox ORP Electrodes
An ORP/Redox electrode featuring a metal ring indicator designed for measuring oxidation/reduction potential; ideal for water and other general applications.
Supplier: METTLER TOLEDO™ 51343205
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Measuring oxidation/reduction potential (ORP)
- Drinking water
- Cooling towers
- Swimming pools
- Potable water supplies
- Waste water
- General lab
Accessories & Care
- Ceramic junction (excluding the InLab Redox Flow with movable glass sleeve junction)
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- InLab Redox Pt805 and InLab Redox Ag805: no electrolyte required and must be paired with InLab Reference (51343190)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
S7 | |
Glass | |
Combination Electrode; Silver ring indicator | |
For measuring the oxidation/reduction power of solution | |
ARGENTHAL with Ag+ trap | |
1mol/L KNO3 | |
0°C to 100°C | |
120 mm | |
12 mm |
Ceramic | |
0°C to 100°C | |
1mol/L KNO3 | |
S7 to BNC (30281915) | |
InLab™ Redox Ag | |
32°F to 212°F | |
4.72 in. | |
0.47 in. |