missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLER TOLEDO™ InLab™ Redox ORP Electrodes
An ORP/Redox electrode featuring a metal ring indicator designed for measuring oxidation/reduction potential; ideal for water and other general applications.
$604.33 - $1204.95
Specifications
Connector Types | S7 |
---|---|
Material | Glass |
For Use With (Application) | For measuring the oxidation/reduction power of solution |
For Use With (Equipment) | S7 to BNC (30281915) |
Catalog Number | Mfr. No. | Model | Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Type | Price | Quantity | ||||
01-911-498
|
METTLER TOLEDO™
51343200 |
InLab™ Redox | Combination Electrode; Platinum ring indicator |
Each for $768.98
|
|
||||
01911517
|
METTLER TOLEDO™
51343205 |
InLab™ Redox Ag | Combination Electrode; Silver ring indicator |
Each for $853.75
|
|
||||
01911516
|
METTLER TOLEDO™
51343204 |
InLab™ Redox Au | Combination Electrode; Gold ring indicator |
Each for $1,083.85
|
|
||||
01911502
|
METTLER TOLEDO™
51343201 |
InLab™ Redox Flow | Combination Electrode; Platinum ring indicator |
Each for $1,204.95
|
|
||||
01911503
|
METTLER TOLEDO™
51343203 |
InLab™ Redox Micro | Combination Electrode; Platinum ring indicator |
Each for $604.33
|
|
||||
01911499
|
METTLER TOLEDO™
51343202 |
InLab™ Redox-L | Combination Electrode; Platinum ring indicator |
Each for $853.75
|
|
||||
01920162
|
METTLER TOLEDO™
59904377 |
InLab™ Redox Pt805 | Half-Cell Electrode; Platinum ring indicator |
Each for $798.64
|
|
||||
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Measuring oxidation/reduction potential (ORP)
- Drinking water
- Cooling towers
- Swimming pools
- Potable water supplies
- Waste water
- General lab
Accessories & Care
- Ceramic junction (excluding the InLab Redox Flow with movable glass sleeve junction)
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- InLab Redox Pt805 and InLab Redox Ag805: no electrolyte required and must be paired with InLab Reference (51343190)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
S7 | |
Glass | |
For measuring the oxidation/reduction power of solution | |
S7 to BNC (30281915) |