Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits
Mettler Toledo™ SevenGo™ perfectION Combined ISE Membrane Modules
PerfectION combined membrane module makes ISE easy
Supplier: Mettler Toledo™ 51344850
Description
- Allows for precise determinations and replaceable membranes for an optimal Ion measurement
- Combined ion-selective electrode
- Polymer membrane
- Lemo connector
Spécifications
Can flow easily through the junction and junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled | |
ISA | |
1.2m | |
Combined ISE Membrane Module | |
0°C to 40°C |
BNC | |
3.9 ft. | |
32° to 104°F | |
perfectION Combined Ca-2 ISE |