missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ SevenGo™ perfectION Combined ISE Membrane Modules

PerfectION combined membrane module makes ISE easy

$714.14 - $2092.31

Specifications

Applications Can flow easily through the junction and junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled
Filling Solution Type ISA
Temperature (English) Operating 32° to 104°F
Temperature (Metric) Operating 0°C to 40°C
View More Specs

Products 3
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
01-928-258
View Documents
Mettler Toledo™
51344821
PerfectION Combined Ksuperscript+ ISE
Each for $2,092.31
 
01-928-264
View Documents
Mettler Toledo™
51344852
PerfectION Combined NO-3 ISE
Each for $777.63
 
01-928-262
View Documents
Mettler Toledo™
51344850
perfectION Combined Ca-2 ISE
Each for $714.14
 
Description

Description

  • Allows for precise determinations and replaceable membranes for an optimal Ion measurement
  • Combined ion-selective electrode
  • Polymer membrane
  • Lemo connector
Specifications

Specifications

Can flow easily through the junction and junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled
ISA
32° to 104°F
0°C to 40°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'