Micro Essential Lab pH Test Paper, in Vial
Immediate, accurate pH test strips cut and packaged in vials
Supplier:  Micro Essential Lab 165B
 View more versions of this product
Catalog No. 14-853-164B
Explore available promotions
Description
Description
For classroom, laboratory and general pH testing applications.
  • General use pH strips
  • Available in normal or jumbo size strips
Specifications
Specifications
1 to 11
Color chart with sharp color changes at the 1, 3, 5, 7, 9 and 11 full pH units
Wide Range pH Test Strip
2 in.
0.2 in.
To provide the convenience of precut paper strips and a cost comparable to two vials of old style litmus paper making them perfect for classroom use
100 Pack
2 x 0.2 in. (5.04 x 0.5 cm)
5.04 cm
0.5 cm
Product Suggestions
Product Suggestions