Micro Essential Lab pH Test Paper, in Vial
Immediate, accurate pH test strips cut and packaged in vials
$57.94 - $66.00
Specifications
For Use With (Application) | To provide the convenience of precut paper strips and a cost comparable to two vials of old style litmus paper making them perfect for classroom use |
---|
Catalog Number | Mfr. No. | Type | Includes | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Includes | Quantity | Price | Quantity | ||||
14-853-165C
|
Micro Essential Lab
4800 |
Jumbo pH Test Paper Strip | Colorimetric chart | 500 Pack |
Pack of 500 Each for $66.00
|
|
||||
14-853-164B
|
Micro Essential Lab
165B |
Wide Range pH Test Strip | Color chart with sharp color changes at the 1, 3, 5, 7, 9 and 11 full pH units | 100 Pack |
Case of 10 Each for $57.94
|
|
||||
22-143875
|
Micro Essential Lab
165A |
Wide Range pH Test Strip | Color chart with sharp color changes at the 2, 4, 6, 8 and 10 full pH unit | 100 Pack |
Pack of 10 Each for $57.94
|
|
||||
Description
For classroom, laboratory and general pH testing applications.
- General use pH strips
- Available in normal or jumbo size strips
Specifications
To provide the convenience of precut paper strips and a cost comparable to two vials of old style litmus paper making them perfect for classroom use |