FEATHER Micro Ophthalmic Scalpel
FEATHER Micro Ophthalmic Scalpels provide ultimate cutting consistency and sharpness. These scalpels are available in a diverse range of shapes and sizes suitable for a variety of surgical ophthalmology applications.
Supplier:  FEATHER 715
 View more versions of this product
Catalog No. 08-924-20
Explore available promotions
Description
Description
  • Made in Japan
  • Handle color-coded for easy identification of blade shape and designed with optimum weight and shape to fit easily in the hand
  • The knife is housed in a heat-resistant plastic case and can be reused after being washed and autoclave sterilized while still in the case
  • Individually packed and sterilized
  • FEATHER Micro Ophthalmic range includes a variety of blade shapes:
    • Incision Knife: Features straight and ultra-sharp blade precisely honed in micron units for optimum use in semi-laminar incisions and centesis incisions of the sclera and cornea
    • Groove Knife: Features a stainless steel guard to assure a consistent cutting depth for incisions in the sclera and cornea while also preventing obstruction of the viewing field during incision
    • Slit Knife: Used for sclerocornea centesis incisions during cataract surgery.
    • Implant Knife: Used to widen incision wound opening for intraocular lens implantation with trapezoidal shape blade tip to enable safe use
    • Round Tunnel Knife: Features a sharp round blade edge beveled on both sides, perfect for creating scleral tunnel flaps
    • Crescent Knife: Features straight sides with a round tip optimal for creating scleral tunnel flaps.
Specifications
Specifications
Stainless Steel
# 15
Sterile
Autoclavable
5/Pk.
Incision Knife
Reusable
ISO13485:2016
Aluminum