Ansell MICROFLEX™ MidKnight™ Black MK-296 Nitrile Gloves
Powder free, nitrile examination gloves with distinct black color hides stains and provides contrast for improved visibility.
$19.72 - $32.68
Specifications
Certifications/Compliance ASTM D6319, FDA21 CFR 177-2600, ISO 1119
Certifications/Compliance (Europe) EN 1149-1/2/3, EN 420:2003 + A1:2009
Chemotherapeutic Rated No
Applications Healthcare, Laboratory, Industrial
AQL 1.5
View More Specs
Products 6
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-168-967A
View Documents Promotion Details
Ansell
MK296XS
X-Small
Pack of 100 Each for $32.68
Case of 10 Pack for $326.80
19-168-967B
View Documents Promotion Details
Ansell
MK296S
Small
Pack of 100 Each for $19.72
Case of 10 Pack for $197.20
19-168-967C
View Documents Promotion Details
Ansell
MK296M
Medium
Pack of 100 Each for $19.72
Case of 10 Pack for $197.20
19-168-967D
View Documents Promotion Details
Ansell
MK296L
Large
Pack of 100 Each for $19.72
Case of 10 Pack for $197.20
19-168-967E
View Documents Promotion Details
Ansell
MK296XL
X-Large
Pack of 100 Each for $19.72
Case of 10 Pack for $197.20
19-168-967F
View Documents Promotion Details
Ansell
MK296XXL
2X-Large
Pack of 100 Each for $19.72
Case of 10 Pack for $326.80
Description
Description
  • Distinctive black color conceals dirt, stains and oils
  • Nitrile gloves provide enhanced tactile sensitivity
  • Fully textured gloves provide a secure grip in wet and dry conditions
  • Powder free, ambidextrous, non-sterile, single-use
  • Length: 9.6 in. (24 cm)
  • Tested against both fentanyl and gastric acid
  • Applicable industries include healthcare, life sciences, automotive and machinery/equipment
Specifications
Specifications
ASTM D6319, FDA21 CFR 177-2600, ISO 1119
No
1.5
Single-use
5.9 mil
Yes
4.7 mil
Powder Free
0.12 mm
MICROFLEX™ MidKnight™ Black MK-296
9.6 in.
EN 1149-1/2/3, EN 420:2003 + A1:2009
Healthcare, Laboratory, Industrial
Black
Fully Textured
0.15 mm
Nitrile
0.12 mm
4.7 mil
Yes
27 MPa
24.38 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'