missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences MicroLiter 11 mm Crimp Top Autosampler Vial, Cap, and FEP/Butyl Septa Unassembled Component Kits

MicroLiter vials are cleaned and packaged in a certified Class 10000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials

$49.04 - $84.70

Specifications

Closure Color Silver
Color Clear
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass (Type 1)
Pre-slit No
View More Specs

Products 2
Catalog Number Mfr. No. Marking Spot Price Quantity  
Catalog Number Mfr. No. Marking Spot Price Quantity  
15-111-540
View Documents Promotion Details
DWK Life Sciences
111021A
No
Pack of 100 Each for $84.70
 
15-111-558
View Documents Promotion Details
DWK Life Sciences
W111221A
Yes
Pack of 100 Each for $49.04
 
Description

Description

  • Unassembled kits
  • Multiple configurations available
  • Identical, clean, and dependable, and are certified to ensure optimal performance and reproducible results
  • Critical physical dimensions are fully tested, as well as chemical cleanliness testing to ensure that trace levels of residues are not present
Specifications

Specifications

Silver
12 mm
No
FEP, Butyl
MicroLiter
Crimp Cap
11 mm
Clear
Borosilicate Glass (Type 1)
2 mL
100
Aluminum
32 mm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'