missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial Septa
MicroLiter septa for 9mm Screwthread Autosampler Vials are constructed with a wide array of chemical compatibilities to suit a variety of chromatography applications.
Supplier: DWK Life Sciences 090035
Specifications
White, Blue | |
PTFE/Silicone | |
9 mm |
Yes | |
100 | |
9 mm Autosampler Vials - MicroLiter |