Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks
Secure specimens during processing with Micromesh™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks. No need for biopsy pads thanks to the 1676 square 0.36 mm openings of the mesh.
Supplier:  Simport™ Scientific M5075T
 View more versions of this product
Catalog No. 22038521
Explore available promotions
Description
Description
  • In Quickload™ stacks
  • 45° Anterior writing area
  • 1676 square opening (0.36 mm)
  • No biopsy pads needed
  • 1 large 27 mm compartment
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Specifications
2000/Pk.
Yellow
Acetal
Product Suggestions
Product Suggestions