Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks
Secure specimens during processing with Micromesh™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks. No need for biopsy pads thanks to the 1676 square 0.36 mm openings of the mesh.
$877.39 - $877.39
Specifications
Quantity 2000/Pk.
Material Acetal
View More Specs
Products 11
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
22038518
View Documents
Simport™ Scientific
M5072T
White
Case of 2000 Each for $877.39
22038519
View Documents
Simport™ Scientific
M5073T
Pink
Case of 2000 Each for $877.39
22038520
View Documents
Simport™ Scientific
M5074T
Green
Case of 2000 Each for $877.39
22038521
View Documents
Simport™ Scientific
M5075T
Yellow
Case of 2000 Each for $877.39
22038522
View Documents
Simport™ Scientific
M5076T
Blue
Case of 2000 Each for $877.39
22038523
View Documents
Simport™ Scientific
M5077T
Peach
Case of 2000 Each for $877.39
22038524
View Documents
Simport™ Scientific
M5078T
Tan
Case of 2000 Each for $877.39
22038525
View Documents
Simport™ Scientific
M5079T
Gray
Case of 2000 Each for $877.39
22038526
View Documents
Simport™ Scientific
M50710T
Lilac
Case of 2000 Each for $877.39
22038527
View Documents
Simport™ Scientific
M50711T
Orange
Case of 2000 Each for $877.39
22038528
View Documents
Simport™ Scientific
M50712T
Aqua
Case of 2000 Each for $877.39
Description
Description
  • In Quickload™ stacks
  • 45° Anterior writing area
  • 1676 square opening (0.36 mm)
  • No biopsy pads needed
  • 1 large 27 mm compartment
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Specifications
2000/Pk.
Acetal
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'